COMH4211B0E5 Combi-Blocks, Inc.

1-(5-Bromo-2-trifluoromethoxy-phenyl)-ethylamine

  • MDL number MFCD24659027
  • Cas Number : 1337079-47-6
  • Molecular Weight : 284.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3744

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3744
Availability details for
Quantity
  • Availability
  • Add to Cart