COMH4211B0E6 Combi-Blocks, Inc.

5-Nitro-2-(oxetan-3-yloxy)benzonitrile

  • MDL number MFCD31618960
  • Cas Number : 1467036-98-1
  • Molecular Weight : 220.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3745

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3745
Availability details for
Quantity
  • Availability
  • Add to Cart