COMH4211B0E7 Combi-Blocks, Inc.

1-Cyclopropylmethyl-1h-indole-4-carboxylic acid

  • MDL number MFCD21105343
  • Cas Number : 1483738-26-6
  • Molecular Weight : 215.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3746

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3746
Availability details for
Quantity
  • Availability
  • Add to Cart