COMH4211B0E9 Combi-Blocks, Inc.

tert-Butyl 3-(4-formylbenzamido)propanoate

  • MDL number MFCD31614042
  • Cas Number : 1280183-85-8
  • Molecular Weight : 277.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-(4-formylbenzamido)propanoate
Inchi Code 1S/C15H19NO4/c1-15(2,3)20-13(18)8-9-16-14(19)12-6-4-11(10-17)5-7-12/h4-7,10H,8-9H2,1-3H3,(H,16,19)
Inchi Key SBBBJDRQWBKUGQ-UHFFFAOYSA-N

Description

Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3751

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3751
Availability details for
Quantity
  • Availability
  • Add to Cart