COMH4211B0EA Combi-Blocks, Inc.

2-((Tetrahydrofuran-3-yl)oxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

  • MDL number MFCD31613968
  • Molecular Weight : 315.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3754

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3754
Availability details for
Quantity
  • Availability
  • Add to Cart