COMH4211B0ED Combi-Blocks, Inc.

4-Bromo-n-(cyclopropylmethyl)-3,5-difluoroaniline

  • MDL number MFCD31381718
  • Cas Number : 1875199-07-7
  • Molecular Weight : 262.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-N-(cyclopropylmethyl)-3,5-difluoroaniline
Inchi Code 1S/C10H10BrF2N/c11-10-8(12)3-7(4-9(10)13)14-5-6-1-2-6/h3-4,6,14H,1-2,5H2
Inchi Key JVAIFJODXIHCRR-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3757

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3757
Availability details for
Quantity
  • Availability
  • Add to Cart