COMH4211B0EE Combi-Blocks, Inc.

(5-Bromo-4-methyl-pyridin-2-yl)-piperidin-1-yl-methanone

  • MDL number MFCD31613901
  • Cas Number : 2270907-19-0
  • Molecular Weight : 283.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3758

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3758
Availability details for
Quantity
  • Availability
  • Add to Cart