COMH4211B0F0 Combi-Blocks, Inc.

Trans toluene-4-sulfonic acid 3-(4-tert-butoxycarbonylamino-cyclohexyl)-propyl ester

  • MDL number MFCD31579414
  • Cas Number : 2270905-33-2
  • Molecular Weight : 411.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3767

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3767
Availability details for
Quantity
  • Availability
  • Add to Cart