COMH4211B0F2 Combi-Blocks, Inc.

Methyl 5-(benzyloxy)-2-hydroxybenzoate

  • MDL number MFCD20441274
  • Cas Number : 61227-22-3
  • Molecular Weight : 258.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3769

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3769
Availability details for
Quantity
  • Availability
  • Add to Cart