COMH4211B0F3 Combi-Blocks, Inc.

2-(4-Bromo-2-fluoro-phenyl)-n-isopropyl-acetamide

  • MDL number MFCD31613957
  • Cas Number : 1789298-27-6
  • Molecular Weight : 274.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3770

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3770
Availability details for
Quantity
  • Availability
  • Add to Cart