COMH4211B0F4 Combi-Blocks, Inc.

[2-[(2-Methylphenyl)methoxy]phenyl]methanol

  • MDL number MFCD11149609
  • Cas Number : 478189-91-2
  • Molecular Weight : 228.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3771

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3771
Availability details for
Quantity
  • Availability
  • Add to Cart