COMH4211B0F5 Combi-Blocks, Inc.

Methyl 2-ethenyl-4-methylbenzoate

  • MDL number MFCD31579467
  • Cas Number : 131001-90-6
  • Molecular Weight : 176.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3774

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3774
Availability details for
Quantity
  • Availability
  • Add to Cart