COMH4211B0F6 Combi-Blocks, Inc.

3-Amino-n-cyclopropyl-5-fluorobenzamide

  • MDL number MFCD24125092
  • Cas Number : 1534788-90-3
  • Molecular Weight : 194.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-amino-N-cyclopropyl-5-fluorobenzamide
Inchi Code 1S/C10H11FN2O/c11-7-3-6(4-8(12)5-7)10(14)13-9-1-2-9/h3-5,9H,1-2,12H2,(H,13,14)
Inchi Key LLDQEIGASXUCFO-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3775

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3775
Availability details for
Quantity
  • Availability
  • Add to Cart