COMH4211B0F8 Combi-Blocks, Inc.

3-Fluoro-2-(1h-1,2,4-triazol-1-yl)benzonitrile

  • MDL number MFCD18906812
  • Cas Number : 870562-65-5
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3777

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3777
Availability details for
Quantity
  • Availability
  • Add to Cart