COMH4211B0F9 Combi-Blocks, Inc.

1-(4-Bromo-3-fluorophenyl)propan-1-amine

  • MDL number MFCD18703703
  • Cas Number : 1270343-15-1
  • Molecular Weight : 232.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3778

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3778
Availability details for
Quantity
  • Availability
  • Add to Cart