COMH4211B0FA Combi-Blocks, Inc.

2-(4-Methoxy-benzyloxy)-5-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenylamine

  • MDL number MFCD31583247
  • Molecular Weight : 355.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3779

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3779
Availability details for
Quantity
  • Availability
  • Add to Cart