COMH4211B0FB Combi-Blocks, Inc.

Methyl 3-(3-iodopropyloxy)benzoate

  • MDL number MFCD30603071
  • Cas Number : 917902-98-8
  • Molecular Weight : 320.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3782

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3782
Availability details for
Quantity
  • Availability
  • Add to Cart