COMH4211B0FD Combi-Blocks, Inc.

(5-Bromo-2-methylphenyl)(4-methoxyphenyl)methanone

  • MDL number MFCD21508232
  • Cas Number : 333361-13-0
  • Molecular Weight : 305.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3784

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3784
Availability details for
Quantity
  • Availability
  • Add to Cart