COMH4211B0FE Combi-Blocks, Inc.

Heptanedioic acid 2,5-dioxo-pyrrolidin-1-yl ester ethyl ester

  • MDL number MFCD31381623
  • Cas Number : 1824718-91-3
  • Molecular Weight : 285.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3785

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3785
Availability details for
Quantity
  • Availability
  • Add to Cart