COMH4211B0FF Combi-Blocks, Inc.

2,2,2-Trifluoro-1-(2,3,5-trichlorophenyl)ethanol

  • MDL number MFCD31613794
  • Cas Number : 1416980-68-1
  • Molecular Weight : 279.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3791

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3791
Availability details for
Quantity
  • Availability
  • Add to Cart