COMH4211B101 Combi-Blocks, Inc.

Glipizide ep impurity h

  • MDL number MFCD02153164
  • Cas Number : 10080-05-4
  • Molecular Weight : 325.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3793

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3793
Availability details for
Quantity
  • Availability
  • Add to Cart