COMH4211B103 Combi-Blocks, Inc.

1-(3-Bromophenyl)-4-cyclopropyl-butane-1,3-dione

  • MDL number MFCD30536352
  • Cas Number : 2168704-99-0
  • Molecular Weight : 281.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-bromophenyl)-4-cyclopropylbutane-1,3-dione
Inchi Code 1S/C13H13BrO2/c14-11-3-1-2-10(7-11)13(16)8-12(15)6-9-4-5-9/h1-3,7,9H,4-6,8H2
Inchi Key FADITDOGEKTGNI-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3795

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3795
Availability details for
Quantity
  • Availability
  • Add to Cart