COMH4211B104 Combi-Blocks, Inc.

1-(4-Iodophenyl)-3-methylbutan-1-amine hydrochloride

  • MDL number MFCD31561550
  • Cas Number : 1383798-66-0
  • Molecular Weight : 325.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3797

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3797
Availability details for
Quantity
  • Availability
  • Add to Cart