COMH4211B105 Combi-Blocks, Inc.

5-(4-Methoxyphenyl)-2-methylaniline

  • MDL number MFCD12824077
  • Cas Number : 761440-20-4
  • Molecular Weight : 213.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3799

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3799
Availability details for
Quantity
  • Availability
  • Add to Cart