COMH4211B106 Combi-Blocks, Inc.

5-Fluoro-4'-(trifluoromethyl)biphenyl-2-carbonitrile

  • MDL number MFCD18424931
  • Cas Number : 1261878-50-5
  • Molecular Weight : 265.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3800

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3800
Availability details for
Quantity
  • Availability
  • Add to Cart