COMH4211B108 Combi-Blocks, Inc.

6-[4-(2-Amino-ethyl)-phenoxy]-nicotinamide

  • MDL number MFCD20999587
  • Cas Number : 676492-10-7
  • Molecular Weight : 257.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3808

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3808
Availability details for
Quantity
  • Availability
  • Add to Cart