COMH4211B109 Combi-Blocks, Inc.

[4-(2-Fluoro-benzyloxy)-pyrazol-1-yl]-acetic acid

  • MDL number MFCD31579324
  • Cas Number : 1895857-61-0
  • Molecular Weight : 250.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3809

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3809
Availability details for
Quantity
  • Availability
  • Add to Cart