COMH4211B10A Combi-Blocks, Inc.

4-Amino-n-cyclopropyl-2-(2-methoxy-ethoxy)-benzamide

  • MDL number MFCD31583111
  • Cas Number : 2270907-07-6
  • Molecular Weight : 250.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3810

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3810
Availability details for
Quantity
  • Availability
  • Add to Cart