COMH4211B10B Combi-Blocks, Inc.

2-Methoxy-4-(2,2,2-trifluoro-ethoxy)-benzaldehyde

  • MDL number MFCD31539545
  • Cas Number : 2167780-54-1
  • Molecular Weight : 234.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3812

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3812
Availability details for
Quantity
  • Availability
  • Add to Cart