COMH4211B10D Combi-Blocks, Inc.

4-Bromo-3-methoxy-n-(methylsulfonyl)benzamide

  • MDL number MFCD31557923
  • Cas Number : 1694636-73-1
  • Molecular Weight : 308.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3813

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3813
Availability details for
Quantity
  • Availability
  • Add to Cart