COMH4211B10E Combi-Blocks, Inc.

1-[3-Fluoro-4-(propan-2-yloxy)phenyl]ethan-1-amine

  • MDL number MFCD11137246
  • Cas Number : 1019603-18-9
  • Molecular Weight : 197.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3818

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3818
Availability details for
Quantity
  • Availability
  • Add to Cart