COMH4211B10F Combi-Blocks, Inc.

(2-Bromo-4-(trifluoromethyl)phenyl)methanamine hydrochloride

  • MDL number MFCD13185406
  • Cas Number : 1214382-77-0
  • Molecular Weight : 290.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3819

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3819
Availability details for
Quantity
  • Availability
  • Add to Cart