COMH4211B110 Combi-Blocks, Inc.

2-[4-(2-Methoxy-ethoxy)-pyrazol-1-yl]-2-methyl-propionic acid

  • MDL number MFCD31579344
  • Cas Number : 2270911-19-6
  • Molecular Weight : 228.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3820

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3820
Availability details for
Quantity
  • Availability
  • Add to Cart