COMH4211B111 Combi-Blocks, Inc.

4-Bromo-1-(cyclopropylmethyl)pyridin-2(1h)-one

  • MDL number MFCD18374852
  • Cas Number : 1127499-07-3
  • Molecular Weight : 228.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3827

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3827
Availability details for
Quantity
  • Availability
  • Add to Cart