COMH4211B112 Combi-Blocks, Inc.

1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxoboralon-2-yl)phenyl)-1h-1,2,4-trizole

  • MDL number MFCD28678079
  • Cas Number : 1384951-86-3
  • Molecular Weight : 271.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3828

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3828
Availability details for
Quantity
  • Availability
  • Add to Cart