COMH4211B13C Combi-Blocks, Inc.

Tetrahydro-4-(4-iodophenyl)-4-methoxy-2h-pyran

  • MDL number MFCD31579367
  • Cas Number : 894399-72-5
  • Molecular Weight : 318.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3833

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3833
Availability details for
Quantity
  • Availability
  • Add to Cart