COMH4211B13D Combi-Blocks, Inc.

5-Chloro-n-ethylpyrazine-2-carboxamide

  • MDL number MFCD31381727
  • Cas Number : 1642321-68-3
  • Molecular Weight : 185.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3838

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3838
Availability details for
Quantity
  • Availability
  • Add to Cart