COMH4211B13F Combi-Blocks, Inc.

3-Bromo-2-ethoxy-benzylamine hydrochloride

  • MDL number MFCD31539527
  • Cas Number : 2204588-05-4
  • Molecular Weight : 266.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3841

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3841
Availability details for
Quantity
  • Availability
  • Add to Cart