COMH4211B140 Combi-Blocks, Inc.

2-Amino-5-fluoro-n,n-dimethylbenzamide

  • MDL number MFCD11132641
  • Cas Number : 1094352-35-8
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3842

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3842
Availability details for
Quantity
  • Availability
  • Add to Cart