COMH4211B141 Combi-Blocks, Inc.

2-(4-Amino-phenyl)-1-[4-(2,2,2-trifluoro-ethyl)-piperazin-1-yl]-ethanone

  • MDL number MFCD31579555
  • Cas Number : 1292007-41-0
  • Molecular Weight : 301.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3844

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3844
Availability details for
Quantity
  • Availability
  • Add to Cart