COMH4211B142 Combi-Blocks, Inc.

3-Ethynyl-4-methylaniline

  • MDL number MFCD11656005
  • Cas Number : 134690-40-7
  • Molecular Weight : 131.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3845

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3845
Availability details for
Quantity
  • Availability
  • Add to Cart