COMH4211B143 Combi-Blocks, Inc.

2H-Benzimidazol-2-one, 4-bromo-1,3-dihydro-1-(1-methylethyl)-

  • MDL number MFCD28403315
  • Cas Number : 1283176-17-9
  • Molecular Weight : 255.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3850

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3850
Availability details for
Quantity
  • Availability
  • Add to Cart