COMH4211B144 Combi-Blocks, Inc.

4-(2-(2-Bromoethoxy)ethoxy)-2-hydroxybenzaldehyde

  • MDL number MFCD31381630
  • Cas Number : 1942058-14-1
  • Molecular Weight : 289.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3851

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3851
Availability details for
Quantity
  • Availability
  • Add to Cart