COMH4211B149 Combi-Blocks, Inc.

1-(5-Bromo-3-dimethylamino-2-methoxy-phenyl)-ethanone

  • MDL number MFCD31579353
  • Cas Number : 474554-57-9
  • Molecular Weight : 272.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3860

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3860
Availability details for
Quantity
  • Availability
  • Add to Cart