COMH4211B14B Combi-Blocks, Inc.

3-Amino-4-fluoro-n-methoxy-n-methylbenzamide

  • MDL number MFCD12095391
  • Cas Number : 774238-92-5
  • Molecular Weight : 198.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3873

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3873
Availability details for
Quantity
  • Availability
  • Add to Cart