COMH4211B14D Combi-Blocks, Inc.

Diethyl (oxetan-3-ylidenemethyl)phosphonate

  • MDL number MFCD20488600
  • Cas Number : 1221819-45-9
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3875

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3875
Availability details for
Quantity
  • Availability
  • Add to Cart