COMH4211B14E Combi-Blocks, Inc.

Acetic acid, 2-hydrazinyl-2-imino-, ethyl ester

  • MDL number MFCD10699336
  • Cas Number : 53085-26-0
  • Molecular Weight : 131.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3879

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3879
Availability details for
Quantity
  • Availability
  • Add to Cart