COMH4211B14F Combi-Blocks, Inc.

4-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-naphthalene-1-carboxylic acid amide

  • MDL number MFCD24039892
  • Cas Number : 2054338-98-4
  • Molecular Weight : 297.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3881

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3881
Availability details for
Quantity
  • Availability
  • Add to Cart