COMH4211B152 Combi-Blocks, Inc.

2-(4-Iodophenoxy)-4-methoxypyrimidine

  • MDL number MFCD31579365
  • Cas Number : 894400-12-5
  • Molecular Weight : 328.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3890

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3890
Availability details for
Quantity
  • Availability
  • Add to Cart