COMH4211B155 Combi-Blocks, Inc.

2-(2-(2-(3-Aminophenoxyl)ethoxy)ethoxy)ethanol

  • MDL number MFCD31613994
  • Cas Number : 263770-42-9
  • Molecular Weight : 241.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3892

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3892
Availability details for
Quantity
  • Availability
  • Add to Cart